3,4-Dihydroxybenzonitrile, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

538396-25G
SIAL538396-25GEA 124 EUR
SIAL538396-25G
3,4-Dihydroxybenzonitrile, Sigma-Aldrich®
3,4-Dihydroxybenzonitrile
Formula: C₇H₅NO₂
MW: 135.12 g/mol
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 160…163 °C
MDL Number: MFCD00016436
CAS Number: 17345-61-8
EINECS: 241-367-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR