Silver, powder, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

576832-5G
SIAL576832-5GEA 92.5 EUR
SIAL576832-5G
Silver, powder, Sigma-Aldrich®
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR