Isoxazole-5-carbonyl chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

636231-1G
SIAL636231-1GEA 80.5 EUR
SIAL636231-1G
Isoxazole-5-carbonyl chloride, Sigma-Aldrich®
Isoxazole-5-carbonyl chloride
Formula: C₄H₂ClNO₂
MW: 131.52 g/mol
Boiling Pt: 74…76 °C (1013 hPa)
MDL Number: MFCD00067868
CAS Number: 62348-13-4
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR