L(+)-Lactic acid 40% aqueous solution, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

69780-1L 69780-250ML
SIAL69780-1LEA 166 EUR
SIAL69780-1L SIAL69780-250ML
L(+)-Lactic acid 40% aqueous solution, Sigma-Aldrich®
L(+)-Lactic acid
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 235
Melting Pt: 53
Storage Temperature: Refrigerator
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 13,05352

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR