Sodium acetate trihydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Acetic acid sodium salt trihydrate

71188-1KG 71188-5KG 71188-250G
SIAL71188-1KGEA 208 EUR
SIAL71188-1KG SIAL71188-5KG SIAL71188-250G
Sodium acetate trihydrate, Sigma-Aldrich®
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR