Sodium dihydrogen citrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate

71497-1KG 71497-250G
SIAL71497-1KGEA 311 EUR
SIAL71497-1KG SIAL71497-250G
Sodium dihydrogen citrate, Sigma-Aldrich®
Sodium dihydrogen citrate
Formula: C₆H₇NaO₇
MW: 214.11 g/mol
Melting Pt: 209…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00013067
CAS Number: 18996-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR