Sodium salicylate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt

71945-250G 71945-1KG
SIAL71945-250GEA 62 EUR
SIAL71945-250G SIAL71945-1KG
Sodium salicylate, Sigma-Aldrich®
Sodium salicylate
Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Melting Pt: 200
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 12,08819

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR