Sodium sulphate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

71959-1KG 71959-5KG 71959-250G
SIAL71959-1KGEA 211 EUR
SIAL71959-1KG SIAL71959-5KG SIAL71959-250G
Sodium sulphate, Sigma-Aldrich®
Sodium sulphate
Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 888
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR