Neutral red, Sigma-Aldrich®

Supplier: Merck

Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5

M-Clarity™ quality level = MQ100

72210-100G 72210-25G 72210-5G
SIAL72210-100GEA 218 EUR
SIAL72210-100G SIAL72210-25G SIAL72210-5G
Neutral red, Sigma-Aldrich®
Neutral red
Formula: C₁₅H₁₇ClN₄
MW: 288.78 g/mol
Melting Pt: 290
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR