Fluorine doped tin oxide (FTO) glass coated slides
Supplier: Merck
735167-1EA 735140-5EA 735159-5EA 735175-5EA 735183-5EA 735191-1EA 735205-5EA 735213-5EA 735221-1EA 735248-5EA 735256-5EA 735264-1EA
SIAL735167-1EAEA 84 EUR
SIAL735167-1EA SIAL735140-5EA SIAL735159-5EA SIAL735175-5EA SIAL735183-5EA SIAL735191-1EA SIAL735205-5EA SIAL735213-5EA SIAL735221-1EA SIAL735248-5EA SIAL735256-5EA SIAL735264-1EA
Fluorine doped tin oxide (FTO) glass coated slides
Slides Adhesive Slides
FTO glass materials have thermal and heated glass performance properties, while also increasing light transmittance and optimising electrical conductivity. Fluorine doped tin oxide is relatively stable under atmospheric conditions, chemically inert, mechanically hard, high-temperature resistant, has a high tolerance to physical abrasion and is less expensive than indium tin oxide.
  • Electrically conductive for heated and thermal control, dissipation and reduced transmittance
  • Colour neutral glass inhibits reflector colour, increases light transmittance and