N-Acetylglycine(Ac-Gly-OH), Sigma-Aldrich®

Supplier: Merck

Synonyms: Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid

MQ200

A16300-100G A16300-500G A16300-5G
SIALA16300-100GEA 44 EUR
SIALA16300-100G SIALA16300-500G SIALA16300-5G
N-Acetylglycine(Ac-Gly-OH), Sigma-Aldrich®
N-Acetylglycine (Ac-Gly-OH)
Formula: C₄H₇NO₃
MW: 117.1 g/mol
MDL Number: MFCD00004275
CAS Number: 543-24-8
EINECS: 208-839-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR