L(+)-Asparagine monohydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O

MQ200

A4284-100G A4284-1KG A4284-25G
SIALA4284-100GEA 126 EUR
SIALA4284-100G SIALA4284-1KG SIALA4284-25G
L(+)-Asparagine monohydrate, Sigma-Aldrich®
L(+)-Asparagine monohydrate
Formula: C₄H₈N₂O₃·1H₂O
MW: 150.13 g/mol
Melting Pt: 215…217 °C
Density: 1.567 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151038
CAS Number: 5794-13-8
EINECS: 200-735-9
Merck Index: 14,00837

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR