Acetylcholine iodide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2-Acetoxy-N,N,N-trimethylethan-1-aminium iodide, (2-Acetoxyethyl)trimethylammonium iodide

M-Clarity™ quality level = MQ200

A7000-25G A7000-5G
SIALA7000-25GEA 118 EUR
SIALA7000-25G SIALA7000-5G
Acetylcholine iodide, Sigma-Aldrich®
Acetylcholine iodide
Formula: H₃CC(O)O(CH₂)₂N(CH₃)₃I
MW: 273.11 g/mol
Melting Pt: 163…165 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00011815
CAS Number: 2260-50-6
EINECS: 218-862-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR