Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate

A7699-10G A7699-1G A7699-5G
SIALA7699-10GEA 1260 EUR
SIALA7699-10G SIALA7699-1G SIALA7699-5G
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate
Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MW: 551.14 g/mol (anhy)
MDL Number: MFCD00150755
CAS Number: 34369-07-8
EINECS: 213-579-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR