N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES), Sigma-Aldrich®

Supplier: Merck

Synonyms: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid

MQ200

A7949-100G A7949-250G A7949-25G
SIALA7949-100GEA 370 EUR
SIALA7949-100G SIALA7949-250G SIALA7949-25G
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES), Sigma-Aldrich®
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)
Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
MW: 182.2 g/mol
Melting Pt: 270…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00008030
CAS Number: 7365-82-4
EINECS: 230-908-4
Merck Index: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR