Agarose, Sigma-Aldrich®

Supplier: Merck


MQ200

A9539-100G A9539-10G A9539-250G A9539-25G A9539-500G
SIALA9539-100GEA 337 EUR
SIALA9539-100G SIALA9539-10G SIALA9539-250G SIALA9539-25G SIALA9539-500G
Agarose, Sigma-Aldrich®
Agarose
Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR