1-Bromo-3-chloropropane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

B62404-1KG B62404-250G
SIALB62404-1KGEA 117 EUR
SIALB62404-1KG SIALB62404-250G
1-Bromo-3-chloropropane, Sigma-Aldrich®
1-Bromo-3-chloropropane
Formula: BrH₂CCH₂CH₂Cl
MW: 157.44 g/mol
Boiling Pt: 144
Melting Pt: –58 °C
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000998
CAS Number: 109-70-6
EINECS: 203-697-1
UN: 2688
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR