Milk, powder

Supplier: Merck

BCR187-20G
SIALBCR187-20GEA 300 EUR
SIALBCR187-20G
Milk, powder
Milk powder
CAS Number: 999999-99-4
EINECS: 310-127-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR