ω,4-Dibromoacetophenone, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide

MQ200

D38308-100G D38308-10G D38308-50G
SIALD38308-100GEA 137 EUR
SIALD38308-100G SIALD38308-10G SIALD38308-50G
ω,4-Dibromoacetophenone, Sigma-Aldrich®
ω,4-Dibromoacetophenone
Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Boiling Pt: 310
Melting Pt: 107…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000200
CAS Number: 99-73-0
EINECS: 202-783-6
UN: 3261
ADR: 8 II
Merck Index: 13,01412

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR