Dimethyl sulphoxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

MQ200

D4540-100ML D4540-1L D4540-500ML
SIALD4540-100MLEA 58 EUR
SIALD4540-100ML SIALD4540-1L SIALD4540-500ML
Dimethyl sulphoxide, Sigma-Aldrich®
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR