3-Fluorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning


MQ100

F5005-10G F5005-50G
SIALF5005-10GEA 43.2 EUR
SIALF5005-10G SIALF5005-50G
3-Fluorobenzaldehyde, Sigma-Aldrich®
3-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 172…174 °C (1013 hPa)
Density: 1.7156 g/cm³ (20 °C)
Flash Pt: 56
Storage Temperature: Ambient
MDL Number: MFCD00003348
CAS Number: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR