4-Fluoro-7-nitrobenzofurazan, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole, NBD-F

MQ100

F5883-25MG F5883-50MG F5883-5MG
SIALF5883-25MGEA 677 EUR
SIALF5883-25MG SIALF5883-50MG SIALF5883-5MG
4-Fluoro-7-nitrobenzofurazan, Sigma-Aldrich®
4-Fluoro-7-nitrobenzofurazan
Formula: C₆H₂FN₃O₃
MW: 183.1 g/mol
MDL Number: MFCD00010196
CAS Number: 29270-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR