D(+)-Galactosamine hydrochloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: (2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

M-Clarity™ quality level = MQ200

G0500-10MG G0500-25G G0500-5G
SIALG0500-10MGEA 17 EUR
SIALG0500-10MG SIALG0500-25G SIALG0500-5G
D(+)-Galactosamine hydrochloride, Sigma-Aldrich®
D(+)-Galactosamine hydrochloride
Formula: C₆H₁₄ClNO₅
MW: 215.64 g/mol
MDL Number: MFCD00135830
CAS Number: 1772-03-8
EINECS: 217-198-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR