Gentamicin sulphate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

MQ200

G1914-250MG G1914-25G G1914-5G
SIALG1914-250MGEA 72.9 EUR
SIALG1914-250MG SIALG1914-25G SIALG1914-5G
Gentamicin sulphate, Sigma-Aldrich®
Gentamicin sulphate
Melting Pt: 218…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS Number: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR