5-(Hydroxymethyl)-2-furaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 5-Hydroxymethyl-2-furancarbaldehyde, 5-(hydroxymethyl)furan-2-carbaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethylfurfural

MQ200

H40807-1G H40807-5G H40807-250MG
SIALH40807-1GEA 86.2 EUR
SIALH40807-1G SIALH40807-5G SIALH40807-250MG
5-(Hydroxymethyl)-2-furaldehyde, Sigma-Aldrich®
5-(Hydroxymethyl)-2-furaldehyde
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 292
Melting Pt: 27…30 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Refrigerator
MDL Number: MFCD00003234
CAS Number: 67-47-0
EINECS: 200-654-9
Merck Index: 13,04857

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR