Hygromycin B, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

MQ200

H7772-100MG H7772-1G H7772-250MG H7772-50MG
SIALH7772-100MGEA 176 EUR
SIALH7772-100MG SIALH7772-1G SIALH7772-250MG SIALH7772-50MG
Hygromycin B, Sigma-Aldrich®
Hygromycin B
Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR