Indole-5-carboxylic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1H-Indole-5-carboxylic acid

MQ100

I5400-1G I5400-5G
SIALI5400-1GEA 49.2 EUR
SIALI5400-1G SIALI5400-5G
Indole-5-carboxylic acid, Sigma-Aldrich®
Indole-5-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 211…213 °C
Storage Temperature: Freezer
MDL Number: MFCD00005678
CAS Number: 1670-81-1
EINECS: 216-799-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR