4-Methoxy-o-toluidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Methoxy-2-methylaniline, m-Cresidine, 2-Methyl-p-anisidine, 1-Amino-4-methoxy-2-methylbenzene, 2-Amino-5-methoxytoluene, 4-Amino-3-methylanisole

MQ200

M15001-5G
SIALM15001-5GEA 69.3 EUR
SIALM15001-5G
4-Methoxy-o-toluidine, Sigma-Aldrich®
4-Methoxy-o-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 248
Melting Pt: 29…30 °C
Density: 1.065 g/cm³ (20 °C)
MDL Number: MFCD00007735
CAS Number: 102-50-1
EINECS: 203-036-7
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR