Magnesium chloride, Sigma-Aldrich®

Supplier: Merck


MQ300

M4880-100G
SIALM4880-100GEA 155 EUR
SIALM4880-100G
Magnesium chloride, Sigma-Aldrich®
Magnesium chloride
Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 715
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR