MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt), Sigma-Aldrich®

Supplier: Merck

Synonyms: Sodium 2-hydroxy-3-morpholinopropane-1-sulphonate

M-Clarity™ quality level = MQ200

M5914-100G
SIALM5914-100GEA 213 EUR
SIALM5914-100G
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt), Sigma-Aldrich®
MOPSO sodium salt (3-(4-Morpholino)-2-hydroxypropanesulphonic acid sodium salt)
Formula: C₇H₁₄NNaO₅S
MW: 247.2445 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070007
CAS Number: 79803-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR