3-Methylpiperidine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

M73001-100G M73001-5G
SIALM73001-100GEA 71.6 EUR
SIALM73001-100G SIALM73001-5G
3-Methylpiperidine, Sigma-Aldrich®
3-Methylpiperidine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 125
Melting Pt: –24 °C
Density: 0.8635 g/cm³ (0 °C)
MDL Number: MFCD00005994
CAS Number: 626-56-2
EINECS: 210-953-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR