High Temperature Viscosity Standards, Supelco®
Supplier: Merck
N100HT-500ML
SIALN100HT-500MLEA 222 EUR
SIALN100HT-500ML
High Temperature Viscosity Standards, Supelco®
Standards Viscometry Standards
  • Temperature range: 100 to 150 °C
  • Kinematic (cSt), dynamic viscosity (mPa·s) and density (g/ml) given at all temperatures
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
Certifikace: Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation.