Okadaic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin, OA

M-Clarity™ quality level = MQ200

O9381-.1MG O9381-25UG
SIALO9381-.1MGEA 877 EUR
SIALO9381-.1MG SIALO9381-25UG
Okadaic acid, Sigma-Aldrich®
Okadaic acid
Formula: C₄₄H₆₈O₁₃
MW: 805.02 g/mol
MDL Number: MFCD00083455
CAS Number: 78111-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR