Sodium pyruvate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

MQ200

P2256-25G P2256-100G P2256-10MG P2256-500G P2256-5G
SIALP2256-25GEA 49.6 EUR
SIALP2256-25G SIALP2256-100G SIALP2256-10MG SIALP2256-500G SIALP2256-5G
Sodium pyruvate, Sigma-Aldrich®
Sodium pyruvate
Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR