Sodium pyruvate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

MQ200

P5280-25G P5280-100G P5280-500G
SIALP5280-25GEA 75.7 EUR
SIALP5280-25G SIALP5280-100G SIALP5280-500G
Sodium pyruvate, Sigma-Aldrich®
Sodium pyruvate
Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR