Retinoic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin

M-Clarity™ quality level = MQ200

R2625-50MG R2625-100MG R2625-1G R2625-500MG R2625-5G
SIALR2625-50MGEA 61.1 EUR
SIALR2625-50MG SIALR2625-100MG SIALR2625-1G SIALR2625-500MG SIALR2625-5G
Retinoic acid, Sigma-Aldrich®
Retinoic acid
Formula: C₂₀H₂₈O₂
MW: 300.44 g/mol
MDL Number: MFCD00001551
CAS Number: 302-79-4
EINECS: 206-129-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR