Disodium hydrogen phosphate, Sigma-Aldrich®

Supplier: Merck

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

MQ200

S5136-100G S5136-1KG S5136-500G S5136-5KG
SIALS5136-100GEA 43 EUR
SIALS5136-100G SIALS5136-1KG SIALS5136-500G SIALS5136-5KG
Disodium hydrogen phosphate, Sigma-Aldrich®
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR