Sulphachloropyridazine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Amino-N-(6-chloropyridazin-3-yl)benzenesulphonamide

M-Clarity™ quality level = MQ200

S9882-100G
SIALS9882-100GEA 160 EUR
SIALS9882-100G
Sulphachloropyridazine, Sigma-Aldrich®
Sulphachloropyridazine
Formula: C₁₀H₉ClN₄O₂S
MW: 284.7221 g/mol
MDL Number: MFCD00057371
CAS Number: 80-32-0
EINECS: 201-269-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR