Tricine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine

MQ400

T0377-100G T0377-1KG T0377-250G T0377-25G T0377-25KG T0377-5KG
SIALT0377-100GEA 162 EUR
SIALT0377-100G SIALT0377-1KG SIALT0377-250G SIALT0377-25G SIALT0377-25KG SIALT0377-5KG
Tricine, Sigma-Aldrich®
Tricine
Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: 187
Density: 1.523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR