3,3',5,5'-Tetramethylbenzidine (TMB) 98 TLC_ASSAY_METHOD, Sigma-Aldrich®

Supplier: Merck

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

MQ200

T2885-100MG T2885-1G T2885-5G
SIALT2885-100MGEA 49.9 EUR
SIALT2885-100MG SIALT2885-1G SIALT2885-5G
3,3',5,5'-Tetramethylbenzidine (TMB) 98 TLC_ASSAY_METHOD, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidine (TMB)
Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00007748
CAS Number: 54827-17-7
EINECS: 259-364-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR