L(-)-Tryptophan, Sigma-Aldrich®

Supplier: Merck

Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH

MQ200

T8941-100G T8941-10MG T8941-1KG T8941-25G
SIALT8941-100GEA 492 EUR
SIALT8941-100G SIALT8941-10MG SIALT8941-1KG SIALT8941-25G
L(-)-Tryptophan, Sigma-Aldrich®
L(-)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 290
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR