Tricine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine

MQ400

T9784-100G T9784-1KG T9784-250G T9784-25G
SIALT9784-100GEA 207 EUR
SIALT9784-100G SIALT9784-1KG SIALT9784-250G SIALT9784-25G
Tricine, Sigma-Aldrich®
Tricine
Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: 187
Density: 1.523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR