Vanillin, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

M-Clarity™ quality level = MQ300

W310700-10KG-K W310700-1KG-K W310700-25KG-K
SIALW310700-10KG-KEA 1060 EUR
SIALW310700-10KG-K SIALW310700-1KG-K SIALW310700-25KG-K
Vanillin, Sigma-Aldrich®
Vanillin
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 285
Melting Pt: ∼82 °C
Density: 1.056 g/cm³ (20 °C)
Flash Pt: 153
Storage Temperature: Ambient
MDL Number: MFCD00006942
CAS Number: 121-33-5
EINECS: 204-465-2
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=535f9a4c80d51d7343126ae5e79c6c83b3beefa7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR