Coating thickness testers, PosiTector® 6000
Supplier: Industrial Physics
LD6107 LD6099 LD6100 LD6101 LD6102 LD6104 LD6058 LD6106 LD6024 LD6108 LD6109 LD6105 LD6029 LD6023 LD6022 LD6020 LD6018 LD6017 LD6057 LD6027 LD6034 LD6028 LD6019 LD6025 LD6026 LD6014 LD6021 LD6033
SIMELD6107EA 1030 EUR
SIMELD6107 SIMELD6099 SIMELD6100 SIMELD6101 SIMELD6102 SIMELD6104 649-0035 SIMELD6106 SIMELD6024 SIMELD6108 SIMELD6109 SIMELD6105 SIMELD6029 SIMELD6023 SIMELD6022 SIMELD6020 SIMELD6018 SIMELD6017 649-0034 SIMELD6027 SIMELD6034 SIMELD6028 SIMELD6019 SIMELD6025 SIMELD6026 SIMELD6014 SIMELD6021 SIMELD6033
Coating thickness testers, PosiTector® 6000
Coating Testers
This coating thickness gauge is a fully electronic apparatus, that uses magnetic and eddy current principles, to measure coating thickness on both ferrous (Fe) and non ferrous (NFe) metals.
  • Fast measurement for quick inspection
  • Flip display enables right side up viewing
  • USB port for fast connection to a PC with continuous power
Informace pro objednávání: Software Ideal Finish Analysis is available on request. With two user levels Ideal Finish Analysis offers user friendly reporting functions for standard production work, as well as advancedMore Product Information