Tubing, silicone, premier grade, esco®, Sterilin®
Supplier: Sterilin
TSR0550160P
SLINTSR0550160PEA 307 EUR
SLINTSR0550160P
Tubing, silicone, premier grade, esco®, Sterilin®
Tubing Transfer Tubing
Platinum cured superior silicone tubing for medical, pharmaceutical and food applications.
  • Superior clarity and smoother inner surface resulting in less protein binding
  • No contaminating by-products formed during curing (peroxide residues)
  • Batch code laser-etched along full length of tubing for full traceability
  • Suitable for use with peristaltic pumps
  • Suitable for autoclaving or dry heat sterilisation
  • Manufactured under clean room (Class 7 ISO 14644) conditions
Certifikace: USP Class VI, FDA 177.2600 Food Contact, ISO 10993-1.1992, EuropeanMore Product Information