Visible storage cabinet 550, Sovella
Supplier: TRESTON
551-3
SOVE551-3EA 0 EUR
SOVE551-3
Visible storage cabinet 550, Sovella
Furniture Storage Cabinets
Cabinets, which combine a stylish industrial design with practical accessories to provide flexible and efficient small parts storage. The 550 cabinet is designed with strong, dark grey (RAL 7016) polypropylene frame and galvanised steel shelves and ideal for small drawers of crystal clear polystyrene sit securely within the framework.
  • Self adhesive labels are provided for each drawer
The frame is made from polypropylene and the shelves from galvanized steel.