Cysteaminium chloride

Supplier: Spectrum Chemical

Synonyms: 2-Aminoethanethiol hydrochloride, Cysteamine Hydrochloride, 2-Mercaptoethylamine hydrochloride, 2-MEA, β-Mercaptoethylamine hydrochloride, Mercaptoethylamine-HCl, H-Cys-NH2·HCl

C2722-50KGBL C2722-100GM
SPCMC2722-50KGBLEA 0 EUR
SPCMC2722-50KGBL SPCMC2722-100GM
Cysteaminium chloride
Cysteaminium chloride
Formula: C₂H₇NS·HCl
MW: 113.61 g/mol
MDL Number: MFCD00012904
CAS Number: 156-57-0
EINECS: 205-858-1

Order Now

Specification Test Results

Assay 98% Min.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR