Calamine 98,0, powder USP

Supplier: Spectrum Chemical

CA115-12KG CA115-2.5KG CA115-45KGBL
SPCMCA115-12KGEA 1450 EUR
SPCMCA115-12KG SPCMCA115-2.5KG SPCMCA115-45KGBL
Calamine 98,0, powder USP
Calamine

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
CAS Number: 8011-96-9

Order Now

Specification Test Results

Assay (ZnO) 98.0 - 100.5%
Microbial Limits To Pass Test
Loss On Ignition max. 2.0 %
Acid-Insoluble Substances max. 2.0 %
Alkaline Substances To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR