EDTA trisodium salt 99

Supplier: Spectrum Chemical

E1046-12KG E1046-22.5KGBL E1046-50KGBL
SPCME1046-12KGEA 3910 EUR
SPCME1046-12KG SPCME1046-22.5KGBL SPCME1046-50KGBL
EDTA trisodium salt 99
EDTA trisodium salt

Ethylenediaminetetraacetic Acid Trisodium Salt is a colourless aminopolycarboxylic acid that is water soluble. It is typically used to dissolve lime scale. It is valuable because of its role as a six toothed ligand and chelating agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₁₀H₁₃N₂Na₃O₈
MW: 358.19 g/mol
Melting Pt: > 300 °C
Density: 0,7215 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 150-38-9
EINECS: 205-758-8
Merck Index: 12,03558

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR