Iron(III) oxide 95, powder, reagent

Supplier: Spectrum Chemical
Warning

Synonyms: Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide

F1035-2.5KG F1035-500GM F1035-50KG F1035-12KG
SPCMF1035-2.5KGEA 494 EUR
SPCMF1035-2.5KG SPCMF1035-500GM SPCMF1035-50KG SPCMF1035-12KG
Iron(III) oxide 95, powder, reagent
Iron (III) oxide

Ferric Oxide, Powder, Reagent is an inorganic compound and is one of the main oxides of iron. It is used as a polishing agent (jeweler's rouge), a pigment and a photocatalyst. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: Fe₂O₃
MW: 159.69 g/mol
Melting Pt: 1538 °C
Density: 5.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR